Would you like to log in again?
J&W VF-624ms Columns, Agilent Technologies
Supplier: Agilent
J&W
CP9102 CP9103 CP9103 CP9107 CP9105 CP9107 CP9109 CP9100 CP9104 CP9106 CP9105 CP9102 CP9109 CP9100 CP9104 CP9106
AGCP9109EA 995.44 USD InStock
CAAGCP9109 CAAGCP9107 AGCP9109 AGCP9106 AGCP9105 AGCP9107 AGCP9102 AGCP9104 AGCP9103 AGCP9100 CAAGCP9105 CAAGCP9106 CAAGCP9103 CAAGCP9104 CAAGCP9102 CAAGCP9100
J&W VF-624ms Columns, Agilent Technologies
Chromatography Columns
The world's first ultra-low-bleed 6% cyanopropyl/phenyl, 94% polydimethylsiloxane GC column, for analysis of volatile compounds.
  • Designed for the analysis of solvents according to EPA Methods 524, 624, and 8260, as well as USP 467
  • VF-1301ms ultra-low-bleed thin-film has a similar selectivity to 624 and is suitable for semivolatile organic solvents, as well as PCBs and pesticides
  • Enhanced selectivity for USP 467 eliminates co-elution of benzene and 1,2-dichloroethane
  • Mid polarity
  • Low bleed
  • 0.15 mm Int.Ø. columns available for high-efficiency GC and GC/MS analyses
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: AT-624, SPB-624, Rxi-624 Sil MS, Rtx-624, PE-624, 007-624, 007-502, ZB-624Plus, ZB-624
VF-624ms (along with VF-1301ms) is the world's first ultralow-bleed mid-polar 6% cyanopropyl/phenyl, 94% polydimethylsiloxane GC column. It eliminates co-elution of benzene and 1,2-dichloroethane and is designed to provide more accurate trace analysis of semivolatile organic solvents according to EPA methods 524, 624, and 8260, as well as USP 467.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty