Would you like to log in again?
QuickPick Traps and filters, Agilent Technologies
Supplier: Agilent
G1544-80530 G1544-60610 G1544-60610 G1544-80530 CP954963 CP954963
AGG1544-80530PK 154.81 USD InStock
AGG1544-80530 CAAGCP954963 CA76508-256 AGG1544-60610 AGCP954963 CAG1544-60610
QuickPick Traps and filters, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Split vent trap for your GC/MS System.
  • GC traps for superior application performance
Agilent provides all the GC Traps needed to ensure each application can be run with highest performance.
All parts are manufactured to Agilent's high quality standards and have been tested for best performance in Agilent instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty