Would you like to log in again?
J&W Intuvo Polyethylene Glycol (WAX) Columns, Agilent Technologies
Supplier: Agilent Technologies
19091F-115-INT 122-3232-INT 19091F-115-INT 122-3232-INT CP97723-INT 123-3234-INT 123-3234-INT CP97723-INT
AG122-3232-INTEA 918 USD InStock
CAAG19091F115I AG122-3232-INT CAAG1223232INT CAAGCP97723INT CAAG1233234INT AG19091F115INT AGCP97723-INT AG123-3234-INT
J&W Intuvo Polyethylene Glycol (WAX) Columns, Agilent Technologies
Chromatography Columns
Advanced range of WAX columns based on the polyethylene glycol (PEG) polymer, for use with the Agilent Intuvo 9000 GC system.
  • General-purpose columns with same selectivity as DB-WAX
  • DB WAX UltraInert – Optimized inertness
  • HP-INNOWax – Highest upper temperature limits of bonded PEG phases
  • DB-WAXetr – Extended temperature range
  • DB-FFAP – Nitroterephthalic acid modified
  • Part of optimized GC flow path with Agilent Ultra Inert Liners, low-bleed septa, certified vials, and gas clean filters
  • For use with the Agilent Intuvo 9000 System
Agilent offers a range of industry-proven WAX columns for use with the Intuvo 9000 GC System: DB-WAX Ultra Inert, HP-INNOWax, DB-WAXetr, and DB-FFAP. Each phase is based on the polyethylene glycol (PEG) polymer; strict control of the crosslinking and deactivation processes results in various unique phase characteristics to meet your specific analytical needs.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty