Would you like to log in again?
Autosampler Needles and Plungers, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CrossLab
G1313-87203 5181-8811 G1313-87202 G4277-80125 RF0052T-S 5190-1571 8010-0367 8002-0005 RF0050T-S 5190-1549 G1811-23200 5190-1477 5182-0830 5190-1461 5190-1538 G7129-87200 5190-1482 G1313-87201 5190-1473 8001-0005 5190-1496 5190-1478 G3160-65306 5181-8811 5190-1582 G1329-80001 5190-1473 G1811-23200 RF0052T 8001-0007 5181-7442 5190-1549 8001-0014 5182-0830 5190-1548 G7129-87201 5190-1582 G7129-87201 G7129-87202 G7129-87200 G3160-65306 RF0052T-S 5190-1548 5190-1477 5181-7442 G1313-87202 RF0050T G1313-87203 5190-1550 5190-1482 5190-1496 5190-1461 RF0050T G4277-80125 5190-1571 5190-1550 G7129-87202 8001-0005 5190-1538 G1329-80001 5190-1581 8002-0005 G1313-87201 8010-0367 RF0050T-S 5190-1581 5190-1478 8001-0014 RF0052T 8001-0007
AG5190-1571PK 32.35 USD InStock
CAAG5190-1496 CAG1313-87203 CAAG5190-1571 AG5190-1571 AG5190-1496 AGRF0050T-S CAAG8001-0007 CAAG5190-1538 CAG1329-80001 CAAG8001-0005 AG8001-0005 AG5190-1538 AG8001-0007 CAAGRF0050T-S AG5190-1482 CAAGRF0050T AGG4277-80125 AG5182-0830 CAAG5190-1582 CAAG5190-1461 CAAG5190-1581 CAAG5190-1549 CAAG5190-1548 CAAG8001-0014 AGG1329-80001 CAAGG427780125 AG5181-7442 AG5181-8811 AGG1313-87201 AGG1313-87202 AGG1313-87203 AG8002-0005 AGG3160-65306 AG8010-0367 AGRF0050T CAAG5190-1473 AG5190-1473 AG5190-1550 CAAG5190-1550 AGG7129-87202 AG5190-1477 AGG7129-87200 AGG7129-87201 AGRF0052T-S AG5190-1478 CAAGRF0052T-S CAAG5190-1478 CAAG5190-1477 CAG3160-65306 CAAGG181123200 CAAGRF0052T CAAG8002-0005 AG5190-1581 CAAG5182-0830 AG5190-1461 AG5190-1582 CAAG5190-1482 CAAG8010-0367 AGG1811-23200 CAAG5181-7442 AG8001-0014 AG5190-1549 AG5190-1548 CA76508-240 CA76508-242 CAG7129-87202 CAG7129-87200 CAG7129-87201 CAAG5181-8811 AGRF0052T
Autosampler Needles and Plungers, Agilent Technologies
Syringe Needles
Not ready for a new syringe? Try out our replacement needles and plungers.
  • Available for a variety of syringe makes in a choice of PTFE-tipped or metal fitted plunger to meet your application needs
  • Environmentally friendly packaging, with an improved design that reduces waste
  • Fresh needles help to reduce septum coring and keeps your system working at its full potential
  • Help to maintain a chemically inert fluid path of stainless steel, borosilicate Type I glass, or PTFE
Agilent replacement needles and plungers ensure your syringes continue to function at their highest level of performance. Available in various volumes and styles, you'll be sure to find the product that fits your needs, regardless of syringe make and model. Agilent needles and plungers are manufactured with strict tolerances and intense quality standards. This ensures the product performs the way you expect it to, every single time.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty