Would you like to log in again?
CrossLab Sample Loops for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
G1377-87300 G1313-87309 G7129-60300 G5667-81006 5067-4710 G4267-60311 01078-87302 G4226-60415 G4267-60300 G4267-60501 G7129-60300 G7157-60515 G1313-87303 G1329-87302 G4267-60311 5067-4703 G7129-60400 5067-4703 G4267-60301 G1375-87315 G4267-60511 G1377-87310 G5668-60500 G4267-60511 G1313-87303 G1367-87300 G4267-60300 G1313-87309 G4267-60301 G1367-87300 G4226-60310 G4226-60310 G7129-60400 G7167-68900 G5668-60500 G7129-60500 G1375-87315 G1377-87300 G7129-60500 G4267-60500 G1329-87302 G4267-60500 01078-87302 G5667-81006 G1377-87310 G7167-68900 G4226-60415 G4267-60400 5067-4710 G4267-60400 G4267-60501
AGG4267-60501EA 273.15 USD InStock
AGG4267-60501 AGG7129-60300 AGG4267-60500 AGG4267-60301 AGG4267-60300 CA76515-230 AGG1329-87302 AGG4226-60415 CAG1375-87315 AG01078-87302 AGG7129-60500 CAAGG566781006 AG5067-4703 AGG4267-60511 CAG1329-87302 CAAGG137787310 AGG4267-60311 AGG5668-60500 CAG1377-87300 CAAGG426760501 CAAGG426760301 AGG1377-87310 CA5067-4710 CAG4226-60310 CAG7129-60400 CAG4267-60500 CAG4267-60300 AGG7129-60400 AGG4267-60400 CAG1313-87303 CAAGG426760311 CAAGG566860500 CAG1313-87309 CAAGG426760511 AGG4226-60310 AGG7167-68900 AGG1377-87300 AGG5667-81006 CA76508-012 CAAGG716768900 AGG1367-87300 AG5067-4710 CAAG5067-4703 AGG1375-87315 AGG7157-60515 AGG1313-87309 CAG7129-60500 AGG1313-87303 CAG4226-60415 CAG7129-60300 CAG4267-60400
CrossLab Sample Loops for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Sample Loops
A range of high-quality loop capillaries suitable for use with a variety of autosampling equipment.
  • Agilent LC metering device Supplies are designed to operate with your LC instruments to work together seamlessly for maximum efficiency and performance–regardless of application area
  • Supporting your efforts to increase your analytical, instrument, and laboratory efficiency in your everyday work
Agilent manufactures a range of high-quality loop capillaries suitable for use with a variety of autosampling equipment. Loop capacities vary from 8 µl up to 900 µl; an even larger capacity 5 ml model also available.
Agilent offers both stainless steel and PEEK loops for different pressure ranges and applications.
All loops are are manufactured to exacting quality standards and designed to deliver robust, high-precision performance in various Agilent autosamplers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty