Would you like to log in again?
CrossLab VWD Supplies for HPLC, Agilent Technologies
Supplier: Agilent
5062-8522 G1314-60081 G1314-87301 G1314-87302 G1314-87301 G1314-60186 5062-8522 G1314-60182 G1314-60183 G1314-60186 G1314-87302 G1314-60182 G1314-60081 G1314-60187 G1314-60183 G1314-60187
AG5062-8522EA 54.78 USD InStock
CAAGG131460182 CAAGG131460183 CAAGG131460186 AG5062-8522 CAAGG131460187 CAG1314-87301 CAG1314-87302 CAAGG131460081 CA76514-664 AGG1314-87301 AGG1314-60187 AGG1314-60186 AGG1314-87302 AGG1314-60183 AGG1314-60182 AGG1314-60081
CrossLab VWD Supplies for HPLC, Agilent Technologies
Tubing Chromatography Tubing
High-quality flow cells and repair kits for Agilent Variable Wavelength HPLC detectors.
  • Designed for highest chromatographic performance
The specific repair kit for each flow cell includes all relevant supplies required to produce the highest level of performance in all variable wavelength detectors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty