Would you like to log in again?
CrossLab Detector Kits for HPLC, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
G1314-65061 G1314-65061 G4212-68001 G4212-68001
AGG1314-65061KT 232 USD InStock
CAG1314-65061 CAG4212-68001 AGG1314-65061 AGG4212-68001
CrossLab Detector Kits for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Maintenance kits for your VWD and DAD/MWD.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
Agilent offers maintenance kits to ensure the continued performance of your variable wavelength detector (VWD) and diode array detector/multiple wavelength detector (DAD/MWD). VWD kits include windows, gaskets, screw kits and rings necessary for VWD equipment, and options are available for standard, micro, semi-micro, and high-pressure flow cells. DAD/MWD kits provide a range of options for different capacity flow cells and include all necessary ferrules, seals, wrenches, and torque adaptors required to deliver excellent performance in DAD/MWD applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty