Would you like to log in again?
CrossLab Needle and Needle Seats for HPLC, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CrossLab
G1367-87012 G1367-87201 G4267-87210 G1367-87101 G4226-87012 G2260-87101 G4226-87012 G1329-87017 G1377-87201 G1329-87017 G1313-87102 G4267-87201 G5667-81008 G7129-87017 G1367-87201 G7129-87112 G2258-68710 G7137-87201 G7129-87012 G7129-87112 G7137-87012 5188-8052 G7137-60075 G1367-87012 G7157-87201 G1377-87001 G7167-87308 G1329-87103 G4267-87012 G7129-87117 G1329-87103 G5667-87200 G1377-87001 G7137-60075 G1377-87002 G7129-87017 5188-8052 G7129-87117 G1367-87102 G5667-81008 G7157-87101 G7167-68711 G7167-87308 G1377-87000 G7157-87101 G1329-87101 G4267-87020 G1329-87012 G7137-87201 G1367-87202 G2260-87201 G4226-87201 G1367-87101 G1367-87017 G4226-87030 G2260-87101 G1377-87201 G1377-87000 G1329-87012 G2258-87102 G1329-87101 G4267-87012 G4267-87201 G1367-87102 G4226-87201 G4267-87210 G4267-87020 G1377-87002 G7167-68711 G7137-87012 G2258-68710 G4226-87211 G7157-87201 G1367-87202 G4226-87211 G5667-87200 G1313-87102 G2258-87102 G4226-87030 G7129-87012 G2260-87201 G1367-87017
AGG7157-87201EA 333 USD InStock
CAAGG137787000 AGG7157-87201 CAAGG137787001 CAG1367-87012 CAAGG426787210 CAAGG137787002 AGG5667-87200 AGG7137-87201 CAG1367-87017 CAG2260-87201 AGG4226-87211 AGG4226-87012 AGG1329-87103 CAAGG715787101 AGG4267-87020 AGG1329-87101 AGG1377-87001 AGG1377-87002 AGG1377-87201 AGG1367-87202 AGG1367-87201 CA76515-228 CAG7129-87117 CAAGG713787201 AGG1377-87000 CAAGG712987112 CAG1329-87101 AGG7129-87117 CAG4267-87201 AGG7129-87112 CAG1367-87202 AGG4267-87210 AGG4267-87012 CAAGG422687030 AGG7167-68711 CAG4226-87012 AGG2260-87201 CAG5667-87200 CAAG5188-8052 AGG1367-87017 AGG1367-87012 CAG1313-87102 CAG2260-87101 CAG7129-87012 CAG1329-87017 AGG7157-87101 CAG4267-87012 AGG4226-87030 AGG4267-87201 CAG7167-68711 AGG7167-87308 CAG1329-87012 CAAGG715787201 AGG2258-68710 AGG5667-81008 CAG4226-87201 CA76508-254 CAAGG225887102 AGG1367-87102 AGG1367-87101 CAG7129-87017 CAAGG716787308 CAG1367-87102 AGG7129-87017 AGG7137-60075 CAG1377-87201 AGG7137-87012 AGG7129-87012 AGG4226-87201 AGG1329-87017 CAAGG422687211 AGG2258-87102 AGG2260-87101 AGG1329-87012 CAAGG225868710 AGG1313-87102 AG5188-8052 CAAGG713787012 CAAGG132987103 CAG5667-81008 CAAGG713760075 CAAGG426787020
CrossLab Needle and Needle Seats for HPLC, Agilent Technologies
Syringe Needles
A comprehensive range of needles and needle seats for HPLC samplers.
  • The needle should be replaced when it becomes bent, burred, or blunt, or when it is leaking or plugged
  • Suspect a leak if you notice a trail of buffer crystals on the needle seat
  • If the sample contains particles, the needle seat can become blocked, as this is the first restriction that the sample experiences
  • If this occurs, try backflushing the needle seat capillary
Agilent provides a comprehensive range of needles and needle seats to equip all different HPLC samplers. Specific designs are available to achieve best performance for any application area, supporting your efforts to significantly increase analytical, instrument, and laboratory efficiency in your everyday work. All needles and seats are manufactured to Agilent's highest levels of quality and have been tested in Agilent instruments to ensure they work perfectly with the rest of the system.
All needles and seats are manufactured to Agilent's highest levels of quality and have been tested in Agilent instruments to ensure they work perfectly with the rest of the system.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty