Would you like to log in again?
CrossLab Gas and Liquid Sampling Valve Supplies, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CrossLab
0101-0299 5181-7459 G3440-60600 18900-20250 07675-80050 G3440-60035 G3440-20037 18900-80255 G3440-60336 0101-0303 G3440-60033 G3440-20037 G1540-30024 G3440-60300 0101-0584 G3440-81665 G3440-60310 0101-0303 0101-0584 5181-7459 G1540-30026 G3440-60300 G3480-20002 18900-20281 0101-0299 G3440-60236 G3440-81665 18900-80255 G3440-20035 VLZF1S610 5062-9508 18900-20280 18900-20850 G3440-60600 G3440-20004 G3440-60033 5181-7460 G3440-60234 18900-20250 G3480-20002 G1540-30024 18900-20850 1530-2163 G3440-20033 5062-9510 G3440-60035 1530-2163 5062-9509 G1540-30025 G3440-20035 18900-20280 18900-20300 18900-20281 5062-9511 G3440-20004 18900-20300 5181-7460 5062-9511 18900-21000 G3440-60310 5062-9508 07675-80050 5062-9509 G3440-60234 G3440-60336 G1540-30026 VLZF1S610 G1540-30025 18900-21000 5062-9510 G3440-60236 G3440-20033
AG5062-9510EA 1195 USD InStock
AG5062-9510 CA0101-0584 AG5062-9511 CA5181-7459 AG18900-20280 AGG3440-20033 AGG3440-60336 AGG3440-20035 AG18900-20281 CAAG1890021000 CAAGG344060600 AGG3440-20037 AGG3440-81665 CAAG1890020281 CAAGG344081665 CAAG1890020280 CA0101-0303 CAAG5062-9510 CAAG5062-9511 CAAG5181-7460 CA5062-9508 CAAG5062-9509 CAAGG344020033 AGG3440-60300 CAAGG344020037 AGG3480-20002 CAAGG344020035 CAAGG344060336 AG18900-80255 AGG3440-20004 AG0101-0584 AG18900-21000 AG0101-0303 CA18900-20300 CAAGVLZF1S610 AGG3440-60033 AGG3440-60310 AGVLZF1S610 AGG3440-60035 CAAG1890020850 CAAGG344060300 AGG3440-60234 CAAG1890020250 CAAGG344020004 AGG3440-60236 AG0101-0299 AG5181-7459 AG18900-20300 AG1530-2163 AGG1540-30024 AGG1540-30025 AGG1540-30026 CAAGG344060033 CA0101-0299 CAAGG344060236 CAAGG344060310 CAAGG344060035 CAAGG348020002 AG5181-7460 CA07675-80050 CAAGG344060234 AGG3440-60600 CAAG1890080255 AG07675-80050 AG18900-20250 CAAGG154030024 CAAGG154030025 CAAGG154030026 AG18900-20850 CA1530-2163 AG5062-9508 AG5062-9509
CrossLab Gas and Liquid Sampling Valve Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent’s portfolio of gas and liquid sampling valve supplies are designed for complex applications.
  • Gas sampling valves are available in Hastelloy C, a robust metal alloy ideal for corrosive or polar analytes, or in Nitrionic 60 stainless steel, which is the standard choice for most general applications
  • Gas sampling valves are available as 4, 6, 10, or 14 port valves
  • Sample loops for gas sampling valves are available from 0.025 μl to 10 ml
  • General purpose liquid sampling valves are stainless steel, and available with 0.2 μl, 0.5 μl, or 1 μl
Gas and liquid sampling valves are most frequently used to sample gases or liquids in constantly flowing streams such as those in chemical reactors, chemical transfer lines, natural gas processing, and fermenters.;Sampling valves can also be used to enable multidimensional GC techniques such as backflush, Deans switching, or 2D-GC (GC x GC). However, for nonvalve injections (i.e., made by a typical liquid autosampler), use of capillary flow technology (CFT) is more straightforward. For more challenging valve applications, such as natural gas or transformer oil gas analysis, Agilent offers preconfigured analyzers for optimizing your GC system. Find out more here, or review Agilent’s Analyzer Solutions Guide for the energy and chemical industry.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty