Would you like to log in again?
High Sensitivity Detection Cells, Agilent Technologies
Supplier: Agilent Technologies
G1600-60027 G1600-68714 G1600-60027 G1600-68723 G1600-68715 G1600-68716 G7100-68723 G1600-63200 G1600-63200
101484-644EA 249.23 USD InStock
CAAGG160060027 101484-644 AGG1600-60027 101484-642 CAG1600-63200 101472-726 101484-646 101477-402 AGG1600-63200
High Sensitivity Detection Cells, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
The HSDC is comprised of a fused silica cell body and removable capillaries. The light path through the cell is made from black fused silica which significantly minimizes stray light and defines the aperture for the diode-array spectrometer.
  • Up to 10-fold increase in signal-to-noise
  • Detector linearity beyond 2000 mAU for accurate quantitative analysis
  • Decoupled design allows replaceable capillaries and reduced cost operation
  • Special capillary geometry ensures maintenance of peak symmetry
  • Full diode-array spectral capabilities
  • Design fits all Agilent CE instruments
The reflective interior functions as a 'lightpipe', ensuring almost 100% transmission of light. These properties result in enhanced linearity and unsurpassed spectral fidelity with the diode-array detector. The high linear range allows quantification of both less than 0.1% impurities and the main component in one run. Integral to the HSDC function is maintenance of peak shape. The unique geometry of the capillaries ensure proper coupling with the cell. The end of these capillaries include both a flaring of the inner diameter and beveling of the outer diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty