Would you like to log in again?
CrossLab Uptake PFA Tubing for ICP-MS, Agilent Technologies
Supplier: Agilent
CrossLab
G1820-65105 G1833-65572 G1820-65526 G1820-65105 G1820-65527 G1833-65573 G1833-65572 G1820-65531 G1820-65478 G1820-65478 G1820-65531 G1820-65527 G1820-65526 G1833-65573
AGG1820-65527PK 735.64 USD InStock
CAAGG183365573 CAG1820-65105 CAAGG182065531 AGG1820-65527 AGG1820-65526 CAG1820-65478 CAAGG183365572 AGG1820-65105 AGG1820-65478 AGG1820-65531 CAAGG182065526 CAAGG182065527 AGG1833-65573 AGG1833-65572
CrossLab Uptake PFA Tubing for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
PFA sample uptake tubing with different Int.Ø available to suit your ICP-MS application.
  • PFA tubing is used to introduce sample together with the peristaltic pump tubing to the nebulizer sample inlet
  • It provides chemical resistance, inertness and cleanliness
PFA tubing is used to introduce sample to the peristaltic pump tubing for the sample inlet, and to connect the outlet of this tubing to sample line connector block. This connection grounds the sample introduction tubing, eliminating any build up of electrostatic charge, that could lead to signal instability.
PFA tubing of different Int.Ø's are available so you can select the optimum tubing for your application. Tubing with a PEEK guide to ensure the sample tubing stays in the solution is also available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty