Would you like to log in again?
CrossLab Skimmer Cones for ICP-MS, Agilent Technologies
Supplier: Agilent
CrossLab
G8400-67201 G1833-65497 G3280-67064 G3666-67511 G8400-67202 G3280-67063 G3280-67060 G3270-60106 G1820-65237 G3270-60643 G8400-67201 G3270-65024 G8400-67202 G8400-67200 G3280-67060 G3270-60106 G3666-67511 G1820-65050 G3666-67401 G1820-65237 G1833-65132 G3280-67066 G3666-67421 G3280-67065 G3280-67066 G3270-60643 G3280-67063 G3666-67411 G1833-65497 G3666-67411 G3270-65024 G3280-67065 G3666-67421 G3280-67041 G3280-67041 G3666-67401 G1833-65132 G8400-67200 G1820-65050 G3280-67064
AGG3270-65024EA 933.44 USD InStock
CAAGG182065050 AGG3270-65024 CAAGG182065237 AGG8400-67201 AGG8400-67202 CAAGG366667421 AGG8400-67200 AGG1820-65237 CAAGG366667401 AGG3270-60106 CAAGG328067065 CAAGG328067066 AGG1820-65050 AGG3270-60643 CAG3270-65024 AGG1833-65497 CAAGG328067063 AGG3666-67411 AGG3666-67511 AGG1833-65132 CAAGG328067064 CAG8400-67201 CAAGG183365497 CAG3280-67060 CAG8400-67202 CAAGG327060106 CAAGG327060643 CAG3280-67041 CAG8400-67200 CAAGG366667411 CAAGG366667511 AGG3280-67060 AGG3280-67063 AGG3280-67041 AGG3280-67064 AGG3280-67065 AGG3280-67066 CAG1833-65132 AGG3666-67421 AGG3666-67401
CrossLab Skimmer Cones for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent skimmer cones ensure the performance that single & triple quadrupole ICP-MS systems demand.
  • Nickel skimmer cones are suitable for most common applications and provide the most economical operation
  • Platinum skimmer cones are required for analysis of aggressive acids (especially HF) and when O2/Ar option gas is used for analysis of organic solvents
  • Platinum skimmer with copper base is recommended for the lowest LODs and for higher matrix samples
  • Platinum skimmer with nickel base is recommended for organics analysis
Agilent, nickel or platinum tipped skimmer cones use high-purity materials to minimize background and ensure stability within the plasma. Cones are typically the biggest supplies cost in routine ICP-MS operation. Protect your investment and ensure ease of use by choosing genuine Agilent cones.High-quality interface cones are key to the sensitivity and stability of an ICP-MS. Our skimmer cones are manufactured to stringent specifications and rigorously tested to ensure excellent batch to batch reproducibility. The orifice dimensions and tip geometry are optimized and tightly controlled.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty