Would you like to log in again?
CrossLab LC/MSD Supplies for HPLC, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
9300-2575 8660-0827 05980-60051 G1960-67470 G1958-60137 G1960-80103 G1960-20032 G7604-60000 0100-1855 G1946-67098 G1960-80060 G7604-60000 G1958-67098 9300-2576 1460-2571 G1960-20310 0100-1854 G1946-20301 G1960-67470 9300-2575 0100-1619 G1964-80659 G2571-80103 G1946-68703 G1946-20157 G1946-20157 G2428A G7604-60001 G1946-20213 G1960-80060 1460-2571 G1960-80103 G1946-80054 G2571-80103 9300-2576 5080-5400 G1946-67098 G1960-20032 G2428A G1958-60137 A6103 G1946-80054 G1958-67098 G1946-20301 G1946-68703 G1946-68704 05980-60051 0100-1619 8660-0827 G6301-80004 0100-1854 G6301-80004 A6103 5080-5400 G1946-20213 G1946-68704 0100-1855 G1960-20310 G1947-20029 G1964-80659 G1958-20050 G1947-20029 G1958-20050 G7604-60001
AGG1960-20310EA 136 USD InStock
CA76514-708 CAAGG194680054 AGG1960-20310 AGG1946-20301 AG05980-60051 AGG1960-20032 AGG1960-80060 CA76514-504 CAG1960-80060 CA1460-2571 AGG1960-80103 AGG1946-80054 AGA6103 CAG7604-60000 AGG1958-67098 AG5080-5400 AGG6301-80004 CAAGG194620301 CAG1958-67098 CAG6301-80004 AGG1946-20213 CA8660-0827 AGG1964-80659 CA9300-2575 CA9300-2576 AG8660-0827 AGG7604-60001 AG0100-1855 AGG7604-60000 AG0100-1854 AG0100-1619 CAG1958-60137 AGG1958-20050 CAG1947-20029 CAG1946-68703 AGG1946-67098 CAG2571-80103 AGG2571-80103 CAAGG196020310 CA76514-522 CAAGG196020032 CAAGG194667098 CAG1946-20213 CAG2428A CAAGG196067470 AGG1946-68704 AGG1946-68703 CAAGG194668704 AGG1946-20157 AG9300-2576 AG9300-2575 CA0100-1854 CA0100-1619 AGG1960-67470 AGG1958-60137 CAAGG196080103 AGG1947-20029 CAAGG760460001 CAG1964-80659 AG1460-2571 CAA6103 CAAGG195820050 CAAGG194620157 AGG2428A
CrossLab LC/MSD Supplies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A range of high-quality supplies for LC/MSD to ensure continued high levels of system performance.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
Agilent provides a range of high-quality supplies for LC/MSD to ensure continued high levels of system performance. These supplies include common parts such as ion injectors, capillaries, electron multipliers, and nebulizer needles. LC/MS source supplies ensure maximum source longevity, consistency, and integrity.
Agilent also provides gas purifiers including moisture, hydrocarbon, and universal traps, and a range of specialist tools suitable for LC/MSD calibration, operation, and maintenance. All components and supplies are precision-engineered to Agilent's exacting quality standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty