Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
LC/MS Calibrant Mixes, Agilent Technologies
Supplier: Agilent Technologies
G1969-85020 G1969-85000 G2431A G1969-85010 G1969-85000 G2432A
101472-288EA 309.23 USD InStock
101472-288 CAG1969-85000 101472-766 101401-530 101471-768 AGG1969-85000
LC/MS Calibrant Mixes, Agilent Technologies
Standards Organic Standards
Agilent offers a range of calibrant mixes for your LC/MS source.
  • Typical standards have at least a year of shelf-life
  • Many standards align with a range of applications
Unique formulations that are tested and verified on Agilent instrumentation.
Delivery information: Most standards manufactured with ISO 17025 and Guide 34 certification.
Packaging: Environmentally friendly packaging that is easy-to-open and verify information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty