A range of high-quality tubing made from PEEK, PTFE, and other materials.
- PEEK tubing with ¹/₁₆" (1.6 mm) OD that are flexible and easy to cut to desired lengths, as well as color-coded for easy tracking
- Tubing made of PTFE, PP, or FEP are available for special conditions
- Available accessories include tubing cutters, ferrules, unions, adapters, and more
Agilent provides a range of high-quality tubing made from PEEK, PTFE, and other materials to ensure certain connnections in your LC system. All tubings are manufactured to the highest quality standards to protect the integrity of your results at every step of your LC flow path.