Would you like to log in again?
CrossLab Electron Capture Detector (ECD) for GC, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
G2397-80520 19233-20755 19233-20625 G2397-20540 G1533-80565 19234-60720 19233-20755 19234-60720 5080-8774 G2397-20540 G1533-80565 18713-60050 G2397-80520 19233-20625 18713-60050 5080-8774
AGG2397-20540EA 62.4 USD InStock
CAG1533-80565 CA19233-20625 CA19233-20755 CA19234-60720 AGG2397-20540 CA76514-712 CAG2397-80520 AGG2397-80520 AG19233-20625 AG19233-20755 AG19234-60720 AGG1533-80565 AG18713-60050 CA18713-60050 AG5080-8774 CA76515-258
CrossLab Electron Capture Detector (ECD) for GC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
The Agilent micro ECD has a detection zone volume 10 times smaller than any other ECD and features replicable inlet liner to decreasing column contamination of the cell.
  • Most sensitive on the market
  • Replaceable liner
The Agilent micro ECD is the most sensitive on the market, with a detection zone volume 10 times smaller than any other ECD. The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty