Would you like to log in again?
CrossLab Nebulizers for ICP-MS, Agilent Technologies
Supplier: Agilent
CrossLab
G3139-65108 G3139-65100 G3161-80001 G3139-65108 G3266-80004 G3266-65003 G3266-65003 G3139-65106 G3285-80001 G3161-80001 G3285-80000 G3139-65106 G3266-80005 G1820-65138 G3285-80001 G3285-80002 G3285-80003 G3285-80000 G1820-65138 G3161-80000 G3266-80005 G3139-65102 G3139-65100 G3266-80004 G3139-65102 G3285-80002 G3285-80003 G3161-80000
AGG3139-65102EA 2731.07 USD InStock
AGG3139-65102 AGG3139-65100 AGG3266-80005 CAAGG313965100 AGG1820-65138 CAAGG316180000 CAAGG316180001 AGG3266-80004 CAAGG313965108 CAAGG313965106 AGG3139-65106 AGG3161-80000 AGG3161-80001 AGG3139-65108 CAG3266-80005 CAG1820-65138 AGG3266-65003 CAG3266-80004 AGG3285-80000 CAG3266-65003 AGG3285-80001 CAAGG328580003 AGG3285-80002 AGG3285-80003 CAAGG328580000 CAAGG328580002 CAAGG328580001 CAG3139-65102
CrossLab Nebulizers for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent has MicroMist and inert PFA concentric nebulizers to suit your ICP-MS application needs.
  • Concentric nebulizers are routinely recommended for use with ICP-MS for all but the highest matrix samples which may cause the inner capillary to become clogged by particles or high concentrations of salt
  • Two types of concentric nebulizer are available: Quartz (recommended for trace analysis of clean samples) and PFA (recommended for semi-conductor process chemicals - providing inertness to HF and self-aspiration)
  • High TDS/matrix samples are tolerated by parallel path nebulizers such as the MiraMist available in PEEK or PTFE for inertness to high acid concentration and solvents
  • A full range of connection accessories including U-series and Ezy Fit sample connectors, tubing, O-rings and other fittings are available for all models, plus nebulizer cleaning tools, manufactured to Agilent's high quality standards
  • Ensure the longevity of your nebulizer through regular cleaning with a 5% HNO₃ (v/v) bath (do not use an ultrasonic bath)
  • Replace the nebulizer when the capillary is broken or the nebulizer is damaged
Agilent has a range of nebulizers and supplies to best suit your ICP-MS application needs. From standard glass or inert PFA concentric nebulizers, robust nebulizers for high TDS samples, to low-flow and self-aspirating nebulizers for your ICP-MS.
MicroMist glass concentric nebulizers and a full range of connection accessories including U-series and Ezy Fit sample connectors, tubing, nebulizer cleaning tools, O-rings and other fittings are available for all models.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty