Would you like to log in again?
Speciation Columns for ICP-MS, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
G3154-65001 G3288-80000 G3154-65001 G3154-65002 G3288-80000 G3154-65002 G3268-80001 G3268-80001
AGG3288-80000EA 4217 USD InStock
AGG3288-80000 AGG3154-65001 AGG3154-65002 AGG3268-80001 CAAGG315465002 CAAGG315465001 CAAGG328880000 CAAGG326880001
Speciation Columns for ICP-MS, Agilent Technologies
Chromatography Columns
Agilent Speciation columns ensure excellent peak separation & reproducibility for your applications.
  • Broad range of phases and column configurations including guard columns and high precision speciation columns for the testing of arsenic, chromium and mercury in drinking water, urine, food and other samples
  • All speciation columns are designed to Agilent’s demanding specifications and can help to achieve the market-leading ICP-MS performance that our valued clients have come to expect
  • Ensure speciation analysis results and data are safe, accurate and compliant with regular testing in accordance with local legislation
  • Excellent reproducibility of both the retention time (RT) and concentration helps users determine quantity and chemical species with confidence
  • Excellent peak separation and reproducibility for the most common speciation methods ensure greater productivity
  • With Agilent columns, you can avoid the need for re-analysis of samples
HPLC separation, followed by ICP-MS detection is well established as the analytical method of choice for speciation analysis of many sample types. As a leader in GC and HPLC systems, columns, and ICP-MS instrumentation, Agilent is at the forefront of hyphenated ICP-MS research and development.
For GC or HPLC with ICP-MS to succeed, your chromatographic column must be able to reproducibly separate the various chemical species. Agilent Speciation columns ensure excellent peak separation and reproducibility for the most common speciation methods. Find application specific columns for arsenic (As) or chromium (Cr) in drinking water, urine, foods, and more.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty