Would you like to log in again?
CrossLab Integrated Autosampler (I-AS) Supplies for ICP-MS, Agilent Technologies
Supplier: Agilent
CrossLab
G3160-80066 G3160-65324 G3160-65303 G3160-80065 G3160-65305 G3160-65321 G3160-65307 G3160-80200 G3160-60015 G3160-65326 G3160-80041 G3160-80200 G3160-65325 G3160-65315 G3160-65327 G3160-65328 G3160-65315 G3160-65324 G3160-80065 G3160-65321 G3160-65307 G3160-65304 G3160-60015 G3160-80066 G3160-65305 G3160-65328 G3160-65303 G3160-65320 G3160-80041 G3160-65317 G3160-65327 G3160-65326 G3160-65304 G3160-65320 G3160-65325 G3160-65317
AGG3160-65320EA 132.15 USD InStock
CAAGG316065317 AGG3160-65320 AGG3160-65321 CA76515-268 CAG3160-65305 CA76515-266 CAG3160-65307 AGG3160-65315 AGG3160-65317 CAAGG316065326 CAAGG316065325 CAAGG316065328 CAAGG316065327 CAAGG316060015 CAAGG316065321 CAAGG316065324 CAAGG316080200 CAAGG316080066 CAAGG316065320 CAAGG316080065 CAAGG316080041 AGG3160-60015 AGG3160-80065 AGG3160-80041 AGG3160-80200 AGG3160-80066 CAG3160-65315 AGG3160-65324 AGG3160-65325 AGG3160-65303 AGG3160-65326 AGG3160-65304 AGG3160-65327 AGG3160-65305 AGG3160-65328 AGG3160-65307
CrossLab Integrated Autosampler (I-AS) Supplies for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Integrated autosampler (I-AS) supplies for ICP-MS for ultratrace analysis on Agilent ICP-MS systems.
  • These supplies including replacement tubing kits for in-built peristaltic pump and sample probes, which may be integrated with PFA concentric nebulizers including MicroFlow and C-Flow
  • Covered autosampler for use with the Agilent single and triple quadrupole ICP-MS
  • No metal or drive mechanism moves over sample vials
  • Continuous flowing rinse port for probe wash
  • Choice of sample tray arrangements, small and large sample volumes accommodated
  • Available with 3 × 100 ml standard or wash bottles
  • Fully automated QA/QC control procedures available using the optional Intelligent Sequencing Software
  • Designed for all semiconductor applications and any ultratrace analysis
Agilent offers the fast, flexible, and high-precision integrated autosampler (I-AS) specifically for ultratrace analysis on Agilent ICP-MS systems. The I-AS is ideal for semiconductor applications. In addition, a range of QA/QC control procedures, such as EPA protocols, can be automated using the optional Intelligent Sequencing software.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty