Would you like to log in again?
Nebulizer Supplies for ICP-OES
Supplier: Agilent
CrossLab
G3266-80015 G3266-80020 G3266-80012 G3266-80012 PL0860-0550 G3266-80020 G3266-80015
AGG3266-80012PK 296.23 USD InStock
AGG3266-80012 CAAGG326680012 AGG3266-80015 AGG3266-80020 AGPL0860-0550 CAG3266-80020 CAG3266-80015
Nebulizer Supplies for ICP-OES
Chromatography Supplies Chromatography Spare Parts
Gas/sample line connectors, tubing, and cleaning tools to fit a variety of ICP-OES nebulizers.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
The Agilent ICP-OES series is compatible with a range of accessories and supplies designed to extend the capabilities of the instruments. Agilent has a variety of nebulizer gas/sample connectors, capillary tubing, and cleaning tools to fit a wide variety of nebulizers.
EzyFit and UniFit sample line connectors easily attach sample line tubing to the compatible nebulizer. Sample line connectors are available in 0.5 and 0.75 mm int.Ø. The EzyLok gas inlet connector easily connects the gas line to the nebulizer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty