Would you like to log in again?
CrossLab Drain Tubing for ICP-MS, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CrossLab
G1820-65505 5042-4769 G3280-60077 G1820-65505 G3280-60077 G1820-65515 G3280-60555 G4912-80014 G3160-67000 G1820-65515 G1820-65016 G3280-60555 G3160-67000 G4912-80014 5042-4769 G1820-65016
AGG3160-67000EA 43.35 USD InStock
CAAGG491280014 AGG3160-67000 CAG1820-65515 CAAGG328060555 AG5042-4769 CAAGG182065016 CAAGG316067000 AGG1820-65505 AGG1820-65515 CA5042-4769 AGG4912-80014 AGG1820-65016 CAAGG328060077 CAAGG182065505 AGG3280-60555 AGG3280-60077
CrossLab Drain Tubing for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Drain tubing and fittings for aqueous and organic solvents for all Agilent ICP-MS instruments.
  • Regular inspection of the drain tubing is recommended
  • The drain tubing should be replaced if the tubing has perished, or shows excessive wear or discoloration
  • The regular clear Tygon tubing is suitable for use with aqueous solutuions, including acid digests
  • For compatibility with organic solvents, solvent resistant tubing (ETFE coated Urethane) should be fitted
Drain tubing for aqueous solutions and organic solvents, plus connectors and other fittings are available for all the Agilent ICP-MS instrument models. Drain tubing is used to join the spray chamber waste port to the peristaltic pump tubing to transfer waste solution to the drain tank. The standard tubing is tygon, which is recommended for aqueous samples. For organic solvents, this should be replaced with ETFE coated Urethane tubing that is solvent resistant.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty