Would you like to log in again?
Operating Supplies Kits for ICP-MS, Agilent Technologies
Supplier: Agilent
G3280-67003 G3280-67004 G3280-67085 G3280-67085 G3280-67221 G3280-67221 G3280-67004 G3280-67003
AGG3280-67004KT 12858.88 USD InStock
AGG3280-67004 AGG3280-67003 CAAGG328067003 CAAGG328067004 AGG3280-67085 CAAGG328067221 AGG3280-67221 CAAGG328067085
Operating Supplies Kits for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
ICP-MS operating supplies kits with 12 months consumables, to ensure performance & reduced downtime.
  • Cost-effective genuine Agilent ICP-MS operating supplies kits for 7700, 7800, 7900, 8800, 8900 to maintain accurate analysis
  • Keep your instruments running for up to 12 months without worrying about reordering
  • With configurable kits, you can configure the kits to suit your specific instrument configuration and application;Reduce costs while ensuring long-term confidence in your results
  • Save as much as 15% over the cost of purchasing supplies individually
The Agilent operating supplies kits for ICP-MS includes all the parts and supplies required to support up to 1 year of routine operation, at a lower cost. Agilent configurable parts and consumable kits allow you to configure the contents of the kits for your specific instrument and application. That means you can select the type and quantity of sampling cones, skimmer cones, and other supplies you need, and still benefit from the savings over the cost of purchasing supplies individually. Agilent parts and supplies kits include Agilent certified parts which offer increased performance, reliability, and longevity of your instrument, while reducing instrument downtime. To maintain system performance, replace parts on recommended schedules.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty