Would you like to log in again?
Accessories for Gas Leak Detector, Agilent Technologies
Supplier: Agilent Technologies
G3388-80002 G3388-80002 G3388-80001 G3388-80001
AGG3388-80002EA 344 USD InStock
AGG3388-80002 AGG3388-80001 CAAGG338880002 CAAGG338880001
Accessories for Gas Leak Detector, Agilent Technologies
Gas Detection Systems
Portable, battery-powered electronic leak detector to identify gas leaks in GC systems.
  • Minimum detection limit of 0.0005 ml/min hydrogen and helium
  • Leak detection through differences in thermal conductivity
  • Dual leak indicating LCD and audible alarm system
  • Portable, rechargeable lithium-ion battery holds up to five hours charge
Gas leaks can cause detector noise and baseline instability, shorten column life, and waste expensive carrier gas. The Agilent GC leak detector is an electronic leak detector, which means that there is no danger of contaminating your system as with less expensive liquid-based leak detectors. Always check your valves, fittings, and traps for leaks after every maintenance, and after thermal cycling as these can loosen some types of fittings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty