Would you like to log in again?
CrossLab Ion Lenses for ICP-MS, Agilent Technologies
Supplier: Agilent
CrossLab
G3280-67039 G3280-20646 G8400-60219 G8400-00240 G3666-60303 G1833-65592 G3280-67035 G1833-65593 G8400-60216 G8400-60217 G3666-60503 G3666-60302 G3280-20628 G3280-67038 G3280-20653 G3666-20404 G8400-67003 G3666-67500 G3666-67044 G8400-67047 G3280-01650 G8400-67002 G3666-67400 G3666-20404 G8400-60219 G3666-67500 G1833-65592 G3280-67044 G3666-67044 G3666-67600 G8400-60215 G8400-60218 G3666-67600 G3280-67035 G3280-67039 G3280-60609 G1833-65593 G3280-60615 G3280-60612 G3666-67300 G3666-60403 G8400-67001 G8400-60215 G3280-67037 G3666-60402 G3666-60502 G8400-60216 G3666-60403 G8400-00240 G3666-67038 G8400-67002 G3280-60615 G8400-67003 G8400-67047 G3666-67038 G3280-20628 G3280-67037 G3280-60612 G3280-60613 G3666-60502 G3666-60302 G3666-60503 G3280-67038 G8400-67001 G3666-60402 G3280-67044 G3666-67400 G3280-60613 G3666-60303 G8400-60217 G3280-01650 G3280-60609 G3280-20653 G8400-60218 G3280-20646 G3666-67300
AGG3666-60402EA 515.74 USD InStock
AGG3666-60402 CAAGG328060609 AGG3666-60403 CAAGG366660503 CAAGG366660303 CAAGG366660502 CAAGG366660302 AGG3280-60609 CAAGG366667300 CAAGG366667500 CAAGG840060218 CAAGG840060219 CAAGG328020628 AGG1833-65593 AGG1833-65592 CAAGG328067044 AGG3666-67038 AGG3280-20646 CAG8400-67001 CAAGG183365592 CAAGG183365593 CAAGG366620404 CAAGG328067038 CAAGG328067035 AGG3666-67300 AGG3666-67500 AGG3666-60303 AGG3666-60302 CAAGG328001650 AGG3666-60503 AGG8400-60218 AGG8400-60217 AGG3666-60502 AGG3280-20653 AGG8400-60216 AGG8400-60215 CAAGG366667044 AGG3666-20404 CAAGG366660402 CAAGG366660403 AGG8400-67003 AGG8400-67047 CAAGG366667400 CAAGG328020653 CAAGG366667600 AGG8400-67001 AGG8400-67002 CAAGG328020646 AGG8400-60219 AGG3280-67035 AGG3280-67037 AGG3280-67038 AGG3280-67039 CAAGG840067047 CAAGG840067003 CAAGG840067002 AGG8400-00240 CAAGG328060612 CAAGG840060215 AGG3280-20628 CAAGG328060613 CAAGG840060216 CAAGG328060615 CAAGG366667038 CAAGG840060217 AGG3666-67044 AGG3280-01650 CAAGG840000240 CAG3280-67039 AGG3280-60612 AGG3280-60613 AGG3280-60615 AGG3666-67600 CAG3280-67037 AGG3280-67044 AGG3666-67400
CrossLab Ion Lenses for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Replacement Omega ion extraction lenses for 7700, 7800, 7900, 8800, 8900 ICP-MS with x-lens and s-lens.
  • High transmission off-axis or Omega lens arrangement separates the positively charged ions from the photons and neutral particles
  • Routine x-lens or semiconductor, s-lens, ion extraction lenses are available as individual parts or as an assembly
  • A variety of additional lens components and supplies are available: screw and spacer sets, lens cleaning and polishing kits
  • The extraction lenses are placed just behind the skimmer cone and mounted on the skimmer base
  • They are used to extract positive ions and accelerate them to into the ORS and MS detector
Ion extraction lenses 7700, 7800, 7900, 8800, 8900 ICP-MS perform an essential function of separating the ions from the photons and residual neutral material for the lowest background signal and highest sensitivity with efficient interference removal.;Agilent uses a high transmission off-axis, Omega lens design that is tuned for optimal performance across the mass range.;Replacement Omega ion extraction lens assemblies and individual lens components for replacement and maintenance routine, x-lens, or semiconductor, s-lens, configuration.
Agilent uses a high transmission off-axis, Omega lens design that is tuned for optimal performance across the mass range. Replacement Omega ion extraction lens assemblies and individual lens components for replacement and maintenance routine, x-lens, or semiconductor, s-lens, configuration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty