Would you like to log in again?
CrossLab Bottle Head Assemblies for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
G1361-60022 G7120-60007 5023-3115 G7120-60017 G7161-60042 G7122-60007 G7161-60042 G1361-23205 5023-3115 G1312-68716 G2258-23201 G4220-60017 5004-0005 G4220-60117 G1361-23204 G4220-60007 G2258-23201 G7120-60007 3150-0944 01018-60025 5004-0005 5004-0004 5004-0004 G1311-60003 G1361-23204 G1361-23205 G7120-60017 3150-0944 0905-1516 G4220-60017 5067-5407 G4220-60007 5067-5407 G4220-60117 0905-1516 G1361-60022 01018-60025 G1312-68716 G7122-60007 G1311-60003
AG5067-5407EA 311.6 USD InStock
AG5067-5407 CAG7120-60007 AGG2258-23201 CAAGG136123205 CAAG5023-3115 AG5004-0004 AG5004-0005 AGG7122-60007 CAG7122-60007 AG5023-3115 CAG4220-60007 CA76514-592 AGG4220-60117 AGG4220-60017 CA76508-010 AG0905-1516 AGG1361-23205 AGG1361-23204 CAAGG716160042 CAAG5004-0004 AGG7120-60017 AGG7161-60042 CAAG5004-0005 CA76515-182 AG01018-60025 CAG7120-60017 AGG1311-60003 AGG1312-68716 CA76514-532 AGG4220-60007 CAAGG422060017 AGG1361-60022 CAAGG422060117 CAG2258-23201 CAAGG131268716 AG3150-0944 CA5067-5407 CAG1361-23204 CAAGG136160022 AGG7120-60007
CrossLab Bottle Head Assemblies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
All the parts required to connect solvent bottles to your HPLC instrument.
  • Instrument specific ready-to-use bottle head assemblies available
  • Special ultraclean tubing for MS applications available
Agilent bottle head assemblies contain all the parts that are required to connect solvent bottles to your HPLC instrument. Each assembly contains a solvent inlet filter, tubing, a bottle cap, and fittings.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty