Would you like to log in again?
CrossLab Capillary Kits for HPLC, Agilent Technologies
Supplier: Agilent Technologies
5067-5708 5067-4248 5023-2256 5067-6723 5023-2257 5067-6868 5023-2257 5067-6621 5067-6870 5067-6621 G4309-68715 5067-4769 5065-9972 5067-4769 5065-4495 5067-5708 5023-2256 5067-6870 5067-4730 5023-2258 5067-4248 5067-6724 5067-4601 5067-6868 5067-6867 5067-6724 5067-4767 5067-4767 5067-6867 5067-6869 5023-2258 5067-4601 5067-4730 G4309-68715 5023-2255 5065-4495 5065-9972 5023-2255 5067-6723 5067-6869
AG5065-9972KT 1569 USD InStock
CAAG5067-5708 AG5065-9972 AG5067-5708 CAAG5065-9972 CAAGG430968715 AG5067-6870 AG5067-4730 CAAG5067-4730 CAAG5067-6870 AG5067-4769 AG5067-6869 CAAG5023-2255 CAAG5067-4769 CAAG5067-6869 CAAG5023-2257 CAAG5067-6868 CAAG5067-4601 CAAG5023-2256 CAAG5067-6724 CAAG5067-4767 CAAG5067-6867 CAAG5065-4495 AGG4309-68715 AG5023-2255 AG5023-2258 AG5023-2257 AG5065-4495 AG5023-2256 CAAG5067-6723 CAAG5023-2258 CAAG5067-4248 CAAG5067-6621 AG5067-6621 AG5067-6723 AG5067-4248 AG5067-4601 AG5067-6868 AG5067-6724 AG5067-6867 AG5067-4767
CrossLab Capillary Kits for HPLC, Agilent Technologies
Tubing Chromatography Tubing
System capillaries and fittings for efficienct ramp up of usage of your LC instruments.
  • Carefully selected capillaries dimensions (inner diameter and length) to minimize extra-column volume and optimize separation efficiency
  • Get peace of mind from having everything at hand for all necessary connections on your new systems
  • InfinityLab Quick Connect and Quick Turn fittings in the InfinityLab capillary kits ensure ease-of-use and zero-dead-volume column connections
Agilent InfinityLab capillary kits include a complete set of system capillaries, including InfinityLab Quick Connect and Quick Turn fittings for efficienct ramp up of usage of InfinityLab LC Series instruments. Agilent capillary/fitting starter kits contain the most widely used connection capillaries, fittings, and tools for older LC systems. Dimensions of the capillaries are optimized to minimize extra-column volume to ensure highest separation efficiency.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty