Supplies and sample loops for Agilent ISIS switching valve used with 7700 and 8800 ICP-MS.
- Complete kit of standard consumable parts for ISIS high sample throughput discrete sampling are provided
- Sample loop sizes to suit your application needs are available and customizable to give you complete flexibiliy and optimal sample throughput
- Rotor seals, nuts and ferrules for valves can be provided
Adding the Integrated Sample Introduction System (ISIS) to the 7700 and 8800 ICP-MS dramatically increases sample throughput and reduces maintenance requirements for sample introduction. Designed and manufactured by Agilent, the ISIS features a robust switching valve fitted with sample loop volume to suit your analysis requirements to allow complete flexibility.
The ISIS hardware is built into the ICP-MS instrument chassis and close coupled to increase productivity through high-speed sample uptake and rapid washout. Agilent's ISIS supplies for 7700 and 8800 ICP-MS include switching valve parts, sample loops and all associated tubing and fittings.