Would you like to log in again?
CrossLab Nitrogen Chemiluminescence Detector Supplies for GC, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CrossLab
G6600-80050 G6600-80043 G6600-80044 G6600-60035 G6600-80018 G6600-80043 G6600-80045 G6600-80051 G6600-80044 G6600-80051 G6600-80042 G6600-60038 G6600-60035 G6600-80045 G6600-67008 G6600-80018 G6600-80050 G6600-80063 G6600-80042 G6600-80063 G6600-67008 G6600-60038
AGG6600-60035EA 491 USD InStock
CAG6600-60035 CAG6600-80043 AGG6600-60035 CAG6600-80044 AGG6600-80018 AGG6600-60038 AGG6600-67008 AGG6600-80045 AGG6600-80044 CAAGG660060038 CAAGG660080045 CAAGG660080051 AGG6600-80063 AGG6600-80051 CAAGG660080050 AGG6600-80043 CAAGG660080042 AGG6600-80042 CAAGG660080063 CAAGG660067008 CAAGG660080018 AGG6600-80050
CrossLab Nitrogen Chemiluminescence Detector Supplies for GC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Supplies for 255 Nitrogen Chemiluminescence Detector (NCD). This nitrogen-specific detector is based on a chemiluminescent reaction so complex sample matrices can be analyzed.
  • Complex sample matrices can be analyzed with little or no interference
The Agilent 255 Nitrogen Chemiluminescence Detector (NCD) is a nitrogen-specific detector that produces a linear and equimolar response to nitrogen compounds based on a chemiluminescent reaction of NO with ozone. Even complex sample matrices can be analyzed with little or no interference.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty