Would you like to log in again?
Capillary Cassettes, Agilent Technologies
Supplier: Agilent Technologies
G1600-60002 G7100-60150 G7100-60002 G1600-60002 G7100-68750 G7100-60002 G1600-60150
103577-718EA 492.31 USD InStock
103577-718 AGG7100-60002 CAAGG160060002 CAAGG710060002 AGG1600-60002 101484-556 101443-160
Capillary Cassettes, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Quick-change capillary cassettes that enable capillary exchange in less than a minute.
  • The quick-change cassette allows capillary exchange in less than a minute
  • The cassette and interfaces accept all commercially available capillaries with a 365 µm OD
Agilent provides quick-change capillary cassettes that enable capillary exchange in less than a minute. The color code of the alignment interface must match with the color code of the capillary's built-in alignment stopper.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty