All the parts required to connect solvent bottles to your HPLC instrument.
- Instrument specific ready-to-use bottle head assemblies available
- Special ultraclean tubing for MS applications available
Agilent bottle head assemblies contain all the parts that are required to connect solvent bottles to your HPLC instrument. Each assembly contains a solvent inlet filter, tubing, a bottle cap, and fittings.