Would you like to log in again?
CrossLab Spray Chambers for MP-AES, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
G8000-70007 G8000-70006 G8000-70007 G8000-70006
AGG8000-70007EA 946 USD InStock
CAAGG800070007 CAAGG800070006 AGG8000-70007 AGG8000-70006
CrossLab Spray Chambers for MP-AES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Spray chambers for MP-AES instrument.
  • Double pass glass cyclonic spray chamber is the default configuration recommended for most sample types - it provides improved plasma stability with organic and high TDS samples
  • Single pass glass cyclonic spray chamber is recommended when you need improved signal to noise performance with 'clean' matrices
  • Inert spray chamber is recommended for hydrofluoric (HF) acid digests and high TDS samples - an inert torch and nebulizer is also required for HF acid digests
  • Inert spray chamber features a removable baffle providing two spray chambers in one and improving stability with high TDS samples
  • Inert spray chamber is permenantly treated and never needs to be returned for rejuvenation, ensuring greater uptime and lower operating costs
Agilent has a wide range of spray chambers compatible with your Agilent MP-AES instrument.;The glass cyclonic spray chambers are available in both single and double pass configurations to meet the needs of any sample matrix.
The new Agilent inert spray chamber for HF analysis has a permanently treated sandblasted finish for optimum drainage and continous accurate performance. The inert spray chamber also features a removable baffle allowing for use as a single-pass design for increased sensitivity, or as a double-pass design so you can handle difficult high-solid matrices.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty