Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Application Kits for MP-AES, Agilent Technologies

Supplier: Agilent Technologies

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

G8003-67000 G8003-67000 G8003-67005 G8003-67005
AGG8003-67000KT 1726 USD InStock
AGG8003-67000 CAAGG800367005 CAAGG800367000 AGG8003-67005
Application Kits for MP-AES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
MP-AES application kits for organic solvents, high sensitivity/aqueous samples, and HF acid digests.

  • Application specific sample introduction kits allow you to adapt the instrument to handle challenging samples with different applications
  • Kits are convenient as they ensure you have all the compatible supplies for these challenging applications

You have a choice of convenient kits for handling challenging samples with the Agilent MP-AES instrument. Kits are available for high sensitivity with aqueous samples, organic solvents and an inert kit recommended for use with hydrofluoric acid digests. Each kit includes the recommended nebulizer or torch, spray chamber, and peristaltic pump tubing for that application.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty