Would you like to log in again?
CrossLab Spray Chamber End Caps for ICP-MS, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
G8400-40200 G3280-60008 G3280-60008 G8400-40200
AGG3280-60008EA 331 USD InStock
CAAGG840040200 AGG3280-60008 CAG3280-60008 AGG8400-40200
CrossLab Spray Chamber End Caps for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Spray chamber end cap, nebulizer retaining nut and other supplies for ICP-MS Scott spray chamber.
  • Optional inert PFA Scott type spray chamber provides excellent sensitivity, stability and washout
  • PFA end cap is O-ring free and features a 6 mm nebulizer port and a make up gas port
  • Manufactured from the highest purity grade PFA resin for the lowest blank contribution, ensuring it is suitable for semiconductor applications
The standard quartz sample introduction kit for Agilent ICP-MS instruments features a quartz Scott-type spray chamber offering excellent sensitivity, stability and washout. A separate end cap is fitted to seal the spray chamber and provide a locking fitting to secure the nebulizer in position during operation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty