Would you like to log in again?
Argon Humidifier Supplies for ICP-MS, Agilent Technologies
Supplier: Agilent Technologies
G8400-60700 5190-0148 5190-0148 5069-4496 G8400-60700 5069-4496
AGG8400-60700EA 1936 USD InStock
AGG8400-60700 CAAG5069-4496 AG5190-0148 AG5069-4496 CAAG5190-0148 CAAGG840060700
Argon Humidifier Supplies for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Conquer your challenging high TDS samples with the ICP-MS argon humidifier accessory and supplies.
  • Achieve accurate, reproducible analysis of samples containing elevated levels of total dissolved solids
  • Enables the accurate analysis of challenging sample types that may potentially cause particulate buildup, and eventually block the nebulizer, torch injector or interface cone orifice
  • Reduces frequency of routine maintenance when analyzing samples with elevated TDS
  • Significantly improve long-term stability for analysis of challenging samples
The Agilent argon humidifier accessory is compatible with Agilent 7500, 7700, 7800, 7900, 8800, and 8900 ICP-MS. It reduces instrument drift due to deposits building up in the sample introduction system, typically at the nebulizer, torch injector and interface cone tips, improving long term stability. This accessory is based on hollow fiber membrane tubing, which passes through a bottle containing deionized water. It humidifies the nebulizer gas, reducing desolvation at the nebulizer orifice. This enables analysis of challenging high TDS sample types that could cause salting or particulate buildup.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty