Replacement Omega ion extraction lenses for 7700, 7800, 7900, 8800, 8900 ICP-MS with x-lens and s-lens.
- High transmission off-axis or Omega lens arrangement separates the positively charged ions from the photons and neutral particles
- Routine x-lens or semiconductor, s-lens, ion extraction lenses are available as individual parts or as an assembly
- A variety of additional lens components and supplies are available: screw and spacer sets, lens cleaning and polishing kits
- The extraction lenses are placed just behind the skimmer cone and mounted on the skimmer base
- They are used to extract positive ions and accelerate them to into the ORS and MS detector
Ion extraction lenses 7700, 7800, 7900, 8800, 8900 ICP-MS perform an essential function of separating the ions from the photons and residual neutral material for the lowest background signal and highest sensitivity with efficient interference removal.;Agilent uses a high transmission off-axis, Omega lens design that is tuned for optimal performance across the mass range.;Replacement Omega ion extraction lens assemblies and individual lens components for replacement and maintenance routine, x-lens, or semiconductor, s-lens, configuration.
Agilent uses a high transmission off-axis, Omega lens design that is tuned for optimal performance across the mass range. Replacement Omega ion extraction lens assemblies and individual lens components for replacement and maintenance routine, x-lens, or semiconductor, s-lens, configuration.