Would you like to log in again?
Autosampler Update and Tubing Kits, Agilent Technologies
Supplier: Agilent
G8410-68262 G8410-68262 G8410-68260 G8410-60100 G8410-60100 G8410-68260
AGG8410-68260KT 225.83 USD InStock
CAAGG841068260 CAAGG841068262 AGG8410-68260 AGG8410-60100 AGG8410-68262 CAAGG841060100
Autosampler Update and Tubing Kits, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A range of update kits and optional accessories are available to upgrade sample handling capacity.
  • Upgrade sample handling capacity, giving you flexibility and improving the capabilities of the autosamplers used in your lab
  • Enclose your autosampler to help reduce sample and standard contamination from dust and drafts, and remove corrosive or solvent fumes
  • Update the SPS 4 used with Agilent ICP-MS instruments to enable sampling from deep well 96 well plates (only supported when using MassHunter software with the ICP-MS)
  • Update the autosampler with installation of a carbon fiber probe, to use with strong acid digests or to provide better rigidity in operation
A number of different autosamplers have been used with Agilent AA, ICP-OES, MP-AES and ICP-MS instruments, depending on the specific model. This includes the Agilent SPS 4 used with current instruments, and the SPS 3 or ASX 500 series used with earlier model instruments. Each model autosampler is designed to automate the presentation of liquid samples, drawn from tubes held in racks loaded on the rack location mat, to the instrument for analysis.;A range of optional accessories and update kits are available to upgrade sample handling capacity, giving you flexibility to improve the capabilities of the autosamplers used in your lab.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty