Would you like to log in again?
MSIS and Supplies for ICP-OES, Agilent Technologies
Supplier: Agilent
G8479-60300 G8000-63003 G8479-60100 G8479-60100 7910052900 G8000-63003 G8479-67000 7910052700 G8479-20020 G8479-20020 G8479-67000 3710068900 7910052700 7910052900 3710068900 G8479-60300
AGG8479-60300EA 156.79 USD InStock
CAAG3710068900 AGG8479-60300 CAAGG847960100 AGG8479-60100 CAAG7910052700 AG7910052700 AG3710068900 CAAGG847960300 AG7910052900 AGG8479-67000 CAAGG847920020 AGG8479-20020 CAAGG847967000 CAAGG800063003 AGG8000-63003 CAAG7910052900
MSIS and Supplies for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Simultaneous conventional nebulization and hydride vapor generation for the lowest detection limits.
  • Provides the ability to nebulize a liquid sample and create volatile hydrides simultaneously
  • Improves detection limits on hydride elements: As, Se, Hg
  • Replaces the existing spray chamber and can be used in anyone of three modes: Liquid nebulization only, hydride generation only, or simultaneous solution nebulization and hydride generation
  • Does not require any power or additional gas flows
The Multimode Sample Introduction System (MSIS) provides a convenient route to maximize sensitivity for elements determined by conventional nebulization, and by simultaneous vapor generation of environmentally sensitive elements including As, Se, and Hg with low µg/L detection limits, in a single unit.
Its unique design provides better performance than conventional nebulization through thin film hydride technology, while offering a choice of three modes 1) Conventional nebulization 2) Vapor generation and 3) Simultaneous nebulization/vapor generation), which means that changeover is eliminated and routine and hydride elements can be determined using the same setup.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty