Would you like to log in again?
Tubing Kits for ICP-OES
Supplier: Agilent
CrossLab
9910124500 G8010-60334 9910123600 G8010-60304 9910124300 G3286-80117 9910124300 G8010-60348 G8010-60335 G8010-60334 9910057100 9910124200 G8010-60306 G3286-80118 9910124500 G8010-60307 9910057100 9910097500 G8485-60523 G8010-60303 G8010-60304 9910123900 G8010-60303 G8479-68100 G8010-60307 G8010-60308 G8010-60333 G8010-60308 9910124400 9910097500 9910105400 G8485-60523 G8410-68261 G8010-60335 9910123800 G8479-68100 G8010-60306 9910123900 G8010-60333 9910124400 9910124200 9910105400 9910123800 9910123600 G8010-60348 G3286-80118 G8410-68261 G3286-80117 G8485-60515 G8485-60515
AG9910097500KT 378.27 USD InStock
AG9910097500 CAAG9910124200 AGG8479-68100 CAAG9910105400 CAAG9910124400 AGG8010-60306 AGG8010-60307 AGG8010-60304 CAAGG801060348 CAAGG801060304 AGG8010-60348 AGG8010-60308 CAAGG801060307 AG9910124400 AGG8010-60303 AG9910105400 AG9910124200 CAAG9910123900 CAAGG848560515 AGG3286-80117 AGG3286-80118 CAAG9910097500 CAAGG801060335 CAAGG801060334 AG9910123800 AG9910123600 CAAGG841068261 AGG8010-60335 AGG8485-60523 AGG8010-60333 AGG8010-60334 CAG3286-80117 CAAG9910057100 CAG3286-80118 CAAG9910124300 CAAGG848560523 AGG8410-68261 CAAG9910124500 CAAGG801060333 CAAGG847968100 AG9910057100 AGG8485-60515 AG9910124500 AG9910124300 CAG8010-60308 CAAG9910123800 CAG8010-60306 CAAG9910123600 AG9910123900 CAG8010-60303
Tubing Kits for ICP-OES
Chromatography Supplies Chromatography Spare Parts
Complete tubing kits to match application compatibility for ICP-OES.
  • Convienent tubing kits provide all the compatible tubing needed for your ICP-OES that is compatible with your sample matrix
  • Reduce costs by purchasing a tubing kit vs buying each item individually
Agilent provides convenient tubing kits for handling aqueous samples, semi-volatile organic solvents, volatile organic solvents, ketone-based organic solvents, and aromatic organic solvents for the Agilent ICP-OES instrument. Each kit includes peristaltic pump tubing, drain tubing, waste line, EzyFit connectors, nebulizer capillary tubing, connectors and connecting tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty