Would you like to log in again?
In-line Gas Purifiers, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CrossLab
5182-9401 OT3-4 5060-9084 RMSA-2 MT200-4 5182-9201 HT3-4 MT120-4 5182-9402 MT120-2 HT200-4 RMSN-4 MT200-4 OT1-2 5060-9084 OT-2-SS RMSN-4 RMSH-2-SS OT3-4-SS MT-2-SS OT1-2 MT200-2 RMSH-2-SS RMSH-4-SS HT200-4 BOT-2 HT3-4 HT200-2 HT3-2 5182-9201 RMSH-4-SS RMSA-2 OT3-4-SS 5182-9411 5182-9401 5060-9096 MT120-2 HMT200-4 MT-4-SS 5060-9096 BOT-2 MT120-4 BOT-4 HMT200-4 OT-4-SS MT-2-SS OT3-4 5182-9402 HT200-2 5182-9411 OT-4-SS HT3-2 MT400-2 OT-2-SS 5182-9202 5182-9202 MT400-2 BOT-4 MT-4-SS MT200-2
AGRMSA-2EA 446 USD InStock
AGRMSA-2 CABOT-4 CAAGHT200-2 CAAGMT-4-SS CAAGHT200-4 CAAG5182-9411 AGHT3-4 AGHMT200-4 AG5182-9411 AGOT-2-SS CA5060-9084 AGHT3-2 CABOT-2 AGHT200-4 AGMT-2-SS AG5060-9096 AGHT200-2 AGOT3-4 AG5182-9201 CAAGOT-4-SS AG5182-9202 AG5182-9401 AG5182-9402 AGOT1-2 AGRMSH-4-SS CAAGRMSH-2-SS CAAGHMT200-4 CAAGOT1-2 CAAGMT-2-SS CAAGRMSA-2 AGBOT-2 AG5060-9084 CAOT3-4-SS AGBOT-4 CAAGMT200-2 AGOT-4-SS AGMT120-2 CAAGHT3-2 CAMT120-2 CAAGHT3-4 CAOT3-4 AGMT120-4 CAAGRMSH-4-SS AGMT-4-SS CAMT400-2 AGMT200-2 AGMT200-4 CARMSN-4 CAAG5182-9402 CAAG5182-9202 CAAG5182-9401 AGMT400-2 CAAG5182-9201 CAAGOT-2-SS CAMT200-4 AGRMSN-4 AGRMSH-2-SS AGOT3-4-SS CAAGMT120-4 CA5060-9096
In-line Gas Purifiers, Agilent Technologies
Filters Gas Filters
Inline GC gas purifiers, traps, and filters for removal of oxygen, moisture, and hydrocarbons.
  • Large traps with 250 psig maximum pressure rating and up to 8 liters/min maximum flow
  • Indicating oxygen and moisture purifiers with 100 psig maximum pressure rating
  • Indicating traps change color when adsorption capacity is depleted
  • Moisture or oxygen permeating into gas stream due to a leak is removed by the trap until it expires
Agilent inline gas purifiers (traps or filters) remove common contaminates such as oxygen, moisture, and hydrocarbons from GC carrier and detector gases. Our inline gas purifiers are available in different sizes and configurations such as combination gas traps that remove moisture, oxygen, and/or organics with a single trap. Using oxygen and moisture traps for carrier gas extends column life and protects the instrument.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty