Would you like to log in again?
Sample Compartment Windows for Cary FTIR Systems, Agilent Technologies
Supplier: Agilent
PIKE-160-1127 PIKE-160-1125 PIKE-160-1010 PIKE-160-1015 PIKE-160-1127 PIKE-160-1133 PIKE-160-1133 PIKE-160-1015 PIKE-160-1009 PIKE-160-1125 PIKE-160-1009 PIKE-160-1126 210228390 PIKE-160-1010 210228390 PIKE-160-1126
AG210228390EA 1609.34 USD InStock
CAGPIKE1601133 AG210228390 CAGPIKE1601010 AGPKE-160-1010 AGPKE-160-1133 AGPKE-160-1126 AGPKE-160-1015 AGPKE-160-1125 AGPKE-160-1127 AGPKE-160-1009 CAGPIKE1601009 CAGPIKE1601127 CAAG210228390 CAGPIKE1601125 CAGPIKE1601015 CAGPIKE1601126
Sample Compartment Windows for Cary FTIR Systems, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent offers a wide range of high quality window materials to suit your analytical needs, such as KBr, KRS5 and ZnSe.
  • Ideal for FTIR Spectroscopy
  • Excellent transmission (wavelength range depends on material type)​
  • Good resistance to mechanical shock;Windows are available in standard sizes
  • Windows are individually packaged and silica gel is included to protect windows from exposure to humidity
Important considerations when selecting a window are the wavelength range transmitted, solubility, reactivity, the refractive index, and the mechanical and thermal characteristics.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty