Would you like to log in again?
PLRP-S for Prep to Process, Agilent Technologies
Supplier: Agilent
PL1212-6100 PL1112-6801 PL1212-6100 PL1112-1801 PL1712-6800 PL1212-6801 PL1112-6800 PL1212-1103 PL1212-3703 PL1212-1103 PL1212-3703 PL1112-1802 PL1112-1801 PL1212-6801 PL1112-6800 PL1712-6800 PL1112-6801 PL1112-1802
AGPL1112-1801EA 1100.95 USD InStock
CAAGPL12126801 AGPL1112-1801 AGPL1112-1802 AGPL1712-6800 CAAGPL12121103 AGPL1212-3703 CAAGPL17126800 CAAGPL12123703 CAAGPL11121801 AGPL1112-6800 AGPL1112-6801 AGPL1212-6801 CAAGPL11121802 CAAGPL12126100 AGPL1212-1103 CAAGPL11126801 CAAGPL11126800 AGPL1212-6100
PLRP-S for Prep to Process, Agilent Technologies
Chromatography Columns
Fully porous polystyrene/divinylbenzene media suitable for oligonucleotide and protein purification.
  • Discovery stage to multi-kg cGMP production reduces method development time
  • Chemical stability for separations, optimization, sanitation, and regeneration increases selectivity and column lifetime
  • Single batch packing of multiple columns reduces system downtime and validation costs
The PLRP-S media, rigid poly(styrene/divinylbenzene) particles, are available in a range of pore sizes for small molecule, synthetic biomolecule and macromolecule purification. Their thermal and chemical stability makes them ideal for purifications that require extreme conditions for sample preparation, compound elution, and column regeneration. With a wide choice of pore sizes and extended range of operating conditions, PLRP-S provides more options to achieve the optimum process. Particle sizes range from 3 to 50 μm for scale-up from the μg/mg discovery stage to multi-kg cGMP production. Excellent chemical stability, up to 1 M NaOH, permits sanitation and regeneration that increase column lifetime. PLRP-S media batch sizes of up to 600 L are available, providing single batch packing of multiple columns.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty