Would you like to log in again?
PL Rapide Aqua, Agilent Technologies
Supplier: Agilent
PL1120-3830 PL1120-3830
AGPL1120-3830EA 1673.25 USD InStock
CAAGPL11203830 AGPL1120-3830
PL Rapide Aqua, Agilent Technologies
Chromatography Columns
The PL Rapide Aqua SEC columns offer high speed and resolution on older systems.
  • PL Rapide Aqua SEC columns for high speed and resolution on older systems
  • High-efficiency PL aquagel-OH media combined with wide-diameter hardware
  • Increase overall efficiency by increasing flow rate and reducing run time
The PL Rapide Aqua SEC columns are configured to offer high speed and resolution on older systems by combining high-efficiency PL aquagel-OH media with wide-diameter hardware. By increasing flow rate and reducing run time, overall resolution is improved on systems with high dead volumes and large flow cell detectors.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty