Would you like to log in again?
PL aquagel-OH Analytical, Agilent Technologies
Supplier: Agilent
PL1149-6260 PL1149-6240 PL1549-5800 PL1549-5800 PL1149-6860 PL1120-6830 PL1149-6801 PL1149-6820 PL1149-6801 PL1120-6520 PL1149-6860 PL1149-6850 PL1149-6260 PL1149-6240 PL1149-6820 PL1120-6830 PL1120-6520 PL1149-6850
AGPL1149-6260EA 2230.33 USD InStock
AGPL1149-6260 CAAGPL11496860 CAAGPL11496850 AGPL1149-6240 CAPL1120-6520 AGPL1549-5800 CAPL1120-6830 CAAGPL11496820 CAAGPL11496801 AGPL1149-6801 CAAGPL15495800 AGPL1120-6830 CAAGPL11496260 AGPL1149-6820 CAAGPL11496240 AGPL1149-6850 AGPL1149-6860 AGPL1120-6520
PL aquagel-OH Analytical, Agilent Technologies
Chromatography Columns
PL aquagel-OH SEC columns separate water-soluble polymers and large molecules.
  • Efficient and reliable size exclusion chromatography (SEC) column
  • Meets USP L25, tolerates high buffer concentrations, pH range 2 to 10, temperatures up to 90 °C and organic solvents (up to 50% methanol)
  • Various MW options provide tailored separations
  • Well-tested resistance against interaction with neutral, multiply charged, polyanionic and polycationic samples
The PL aquagel-OH size exclusion chromatography (SEC) column offers efficient and reliable separation of water-soluble polymers and large molecules, such as PEG, polysorbate, starch, dextran, and acrylamide. PL aquagel-OH columns meet USP L25, tolerate high buffer concentrations, pH 2 to 10, temperatures up to 90 °C, and organic solvents (up to 50% methanol). 
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty