Would you like to log in again?
PL HFIPgel, Agilent Technologies
Supplier: Agilent Technologies
PL1514-1900HFIP PL1114-6900HFIP PL1114-6900HFIP PL1514-1900HFIP
AGPL1514-1900HEA 744 USD InStock
AGPL1514-1900H CAGPL11146900H CAGPL15141900H AGPL1114-6900H
PL HFIPgel, Agilent Technologies
Chromatography Columns
PL HFIPgel has been developed to perform reliably in HFIP and other polar fluorinated solvents. 
  • For room-temperature analysis of nylons, polyamides, polyesters, PLLA, and PLGA
  • Reliable results and extended lifetime in HFIP and fluorinated solvents
  • Avoids excessive curvature of calibration curves, dislocations, shoulders, and poor resolution caused by HFIP
  • Optimized separation range delivers high performance with no artifacts
  • Available in 4.6 mm diameters to reduce solvent consumption
PL HFIPgel has been developed to perform reliably in hexafluoroisopropanol, HFIP, and related polar fluorinated solvents. HFIP is a unique solvent that allows ambient temperature GPC of solvent-resistant polymers such as polyesters, polyamides (nylon), polyethylene terephthalate (PET), and poly(lactic-co-glycolic acid) (PLGA). 
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty