Would you like to log in again?
AdvanceBio SEC 2.7 μm, Agilent Technologies
Supplier: Agilent
InfinityLab AdvanceBio
PL1580-3301 PL1180-5301 PL1180-3350 PL1180-1350 PL1180-3350 PL1580-3301 PL1180-1350 PL1180-5301
AGPL1180-3350EA 1823.43 USD InStock
CAAGPL11803350 CAPL1580-3301 CAAGPL11801350 AGPL1180-3350 AGPL1180-5301 AGPL1180-1350 CAPL1180-5301 AGPL1580-3301
AdvanceBio SEC 2.7 μm, Agilent Technologies
Chromatography Columns
Accurate, precise quantitation for mAb aggregation and protein analysis with minimal secondary interactions.
  • High resolution, high-efficiency separation, and accurate quantitation for analysis of aggregates and fragments
  • Reduced secondary interactions with mAb, ADC, and other proteins
  • Assured consistent column performance to eliminate rework
  • Enhanced column lifetime and productivity to meet robust CQA analysis
Agilent AdvanceBio SEC columns ensure easy, fast, and accurate quantitation for mAb aggregation and fragment analysis. AdvanceBio SEC offers two different particle sizes, 2.7 and 1.9 µm. These innovative size exclusion chromatography columns were designed and engineered by Agilent to achieve high resolution and efficiency separation, with a robust and reliable performance. Consistent results are obtained from column to column, batch to batch, and lab to lab, ensuring that methods can be transferred across departments and locations to put an end to uncertainty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty