Atlas mobile carts with heavy-duty leveling casters include two columns with bases, modesty panel and front apron rail with or without front support.
- Sturdy, heavy-duty construction
Carts are available in various sizes and dimensions. 72" wide carts with front support are equipped with a center leg for added support.