Would you like to log in again?
VWR® Phenolic Resin Worksurfaces for Carts, Black
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AMCTWB3572H AMCTWB3172M AMCTWB3548M AMCTWB3160M AMCTWB3136M AMCTWB3130H AMCTWB3530M AMCTWB3160H AMCTWB3172H AMCTWB3548H AMCTWB3572M AMCTWB3148H AMCTWB3536H AMCTWB3130M AMCTWB3560H AMCTWB3536M AMCTWB3530H AMCTWB3136H AMCTWB3560M AMCTWB3148M
AMCTWB3536HEA 1674.04 USD InStock
AMCTWB3536H AMCTWB3548H AMCTWB3548M AMCTWB3136H AMCTWB3148H AMCTWB3172H AMCTWB3560M AMCTWB3572M AMCTWB3130H AMCTWB3136M AMCTWB3148M AMCTWB3530H AMCTWB3536M AMCTWB3572H AMCTWB3160M AMCTWB3172M AMCTWB3130M AMCTWB3560H AMCTWB3530M AMCTWB3160H
VWR® Phenolic Resin Worksurfaces for Carts, Black
Furniture Tables and Workbenches
Phenolic resin worksurfaces are composed of cellulose fiber-reinforced, phenolic resin core with a highly crosslinked, polyurethane, copolymer surface.
  • Each worksurface has three square edges and one edge with a ¹/₈" chamfer
  • For High-Column worksurfaces, two or three cut-outs accomodate 76" high columns, depending on bench width
  • High-Column worksurfaces may be used with sitting/bench height carts when height-adjustment is required
Phenolic resin worksurfaces are chemical-resistant and non-flammable.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty