Would you like to log in again?
BD SafetyGlide™ Syringes for Insulin, TB, and Allergy, BD
Supplier: BD Medical
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
305904 305935 305915 305932 305918 305900 305918 305945 305917 305901 305946 305945 305951 305935 305950 305915 305907 305916 305946 305900 305930 305906 305904 305902 305932 305902 328449 328447 305917 305930 305903 305906 305905 305950 305916 328446 305908 305907 305908 305901 305909 305903 305951 305905
10002-698BX 92.32 USD InStock
10002-698 BD-305904 10002-696 BD305917 CABD305935 CABD305932 BD305918 CABD305916 CABD305917 CABD305915 CABD-305950 BD305930 CABD305918 BD305932 BD305935 BD-305901 BD305915 BD-305903 BD-305916 10002-700 BD305951 CABD305930 CABD305951 CABD305945 CABD305901 BD305906 CABD305946 CABD305902 BD305905 BD305908 CABD305900 BD305907 CABD305905 CABD305906 BD305909 BD-305950 CABD305903 CABD305904 BD305900 CABD305907 CABD305908 BD305946 BD305902 BD305945
BD SafetyGlide™ Syringes for Insulin, TB, and Allergy, BD
Syringes Syringes, Medical
The BD SafetyGlide™ Syringe with Tiny Needle Technology completes the BD SafetyGlide line to meet your Sub-Q and intradermal injection needs
  • BD PrecisionGlide™ needle technology
  • One-handed activation
  • Proven dependability
  • Designed for efficiency
SafetyGlide syringes are designed with a permanently attached needle for minimal deadspace and minimal drug waste. The attached needle with Tiny Needle Technology helps meet Sub-Q and intradermal injection needs of the healthcare workers who administer these types of injections. A rotating safety arm allows for low angle injections.
SafetyGlide needles are designed for patient comfort, featuring one-handed activation. They require virtually no change in technique and require minimal training. Finger stays behind the needle at all times. There is virtually no splatter upon activation. Needles are detachable, allowing clinicians to change needles for aspiration and administration.
The permanently attached needle is designed with low waste space which may result in reduced medication waste.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty