Would you like to log in again?
BD BBL™ CHROMagar™ MRSA II, BD Diagnostics
Supplier: BD Diagnostics
215228 215229
BD215228PK 268.33 USD InStock
BD215228 BD215229
BD BBL™ CHROMagar™ MRSA II, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
BBL™ CHROMagar™ MRSA II is a selective and differential chromogenic medium for the qualitative direct detection of nasal colonization by methicillin-resistant Staphylococcus aureus (MRSA) to aid in the prevention and control of MRSA infections in healthcare settings
The test is performed on anterior nares swab specimens from patients to screen for MRSA colonization. For in vitro diagnostic use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty