Would you like to log in again?
Accessories for Sharps Collectors, BD Medical
Supplier: BD Medical
305495 305485 305020 305447 305447 305485 305451 305017 305497 305494 305550 305017 305448 305588 305497 305420 305570
BD305494CS 96.77 USD InStock
BD305494 BD305020 BD305570 BD305485 BD305495 BD305451 BD305497 BD305588 CABD305497 CABD305485 CABD305447 BD-305550 BD-305420 BD305447 BD-305017 CABD-305017 BD305448
Accessories for Sharps Collectors, BD Medical
Bins Waste Bins
Brackets, cabinets, and supports for BD sharps collectors.
Wire wall brackets are made of powder-coated, formed metal. The Wallmate™ starter kit includes a locking wall bracket, 5.1L (5.4qt.) side entry collector, glove box cabinet bracket (BD305448), and mounting hardware. The security lock consists of a brass padlock and two metal keys with metal ring. The stabilizer is made of high impact polystyrene.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty